Assay Details
| Gene Symbol: MFAP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297709.1 | NP_001284638.1 | 6 - 7 | 480 | 65 | |
| NM_001297710.1 | NP_001284639.1 | 5 - 6 | 444 | 65 | ||
| NR_123734.1 | 6 - 7 | 480 | 65 | |||
| GenBank mRNA | AK299475.1 | - | 6 - 7 | 308 | 65 | |
| AY339060.1 | - | 5 - 6 | 216 | 65 | ||
| BU929788.1 | - | 6 - 7 | 284 | 65 | ||
| BX396041.2 | - | 5 - 6 | 243 | 65 | ||
More Information
Set Membership: |
|

MFAP5
Genomic Map