Assay Details
Gene Symbol: TBCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297738.1 | NP_001284667.1 | 2 - 3 | 267 | 87 | |
NM_004607.2 | NP_004598.1 | 2 - 3 | 267 | 87 | ||
GenBank mRNA | AF038952.1 | - | 2 - 3 | 212 | 87 | |
AK300031.1 | - | 2 - 3 | 189 | 87 | ||
AK312207.1 | - | 2 - 3 | 235 | 87 | ||
BC018210.1 | - | 2 - 3 | 252 | 3861138 | 87 | |
BC105285.1 | - | 2 - 3 | 203 | 6702573 | 87 | |
BT007354.1 | - | 2 - 3 | 163 | 87 | ||
CR542170.1 | - | 2 - 3 | 163 | 87 | ||
CR542186.1 | - | 2 - 3 | 163 | 87 | ||
DC312139.1 | - | 2 - 3 | 267 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001510.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05020692.hg.1 | TC05001510.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05020692.hg.1 | ENST00000306388.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05020692.hg.1 | ENST00000380377.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05020692.hg.1 | ENST00000517679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05020692.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05020692.hg.1 | ENST00000520039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05020692.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05020692.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000520039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000520039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177793.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177793.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177793.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177793.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177793.hg.1 | TC0500011212.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000520039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000520039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177794.hg.1 | TC0500011212.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000520039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000520039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177792.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177792.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177792.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177792.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177792.hg.1 | TC0500011212.hg.1 | Clariom D TC |
