Assay Details
Gene Symbol: TBCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297740.1 | NP_001284669.1 | 2 - 3 | 256 | 124 | |
GenBank mRNA | HY015414.1 | - | 2 - 3 | 233 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000520039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000520039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000520039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000520039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011212.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | ENST00000522370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177791.hg.1 | TC0500011212.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500177783.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177783.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177783.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177783.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177783.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177783.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177783.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177783.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177783.hg.1 | TC0500011212.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000306388.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000380377.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000517679.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000517881.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000518218.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000518338.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | ENST00000520361.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | NM_001297738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | NM_001297740.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | NM_004607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500177789.hg.1 | TC0500011212.hg.1 | Clariom D TC |
