Assay Details
| Gene Symbol: PGAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027016.2 | 1 - 2 | 118 | 58 | ||
| NR_104272.1 | 1 - 2 | 118 | 58 | |||
| GenBank mRNA | CB988453.1 | - | 1 - 2 | 145 | 58 | |
| CX872317.1 | - | 1 - 2 | 46 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nucleoporin 98 | NUP98 |
ADIR2, NUP196, NUP96 |
-10.7kb | Anti-sense | Chr. 11 - 3675010 - 3797792 on NCBI Build GRCh38 |
| Downstream | ras homolog family member G | RHOG | ARHG | 2.8kb | Anti-sense | Chr. 11 - 3826978 - 3840983 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

PGAP2
Genomic Map