Assay Details
Gene Symbol: SCHLAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104321.1 | 2 - 3 | 440 | 71 | ||
GenBank mRNA | JX117420.1 | - | 2 - 3 | 440 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
CWC22 homolog, spliceosome-associated protein |
CWC22 |
EIF4GL, NCM, fSAPb |
-717.3kb | Anti-sense | Chr. 2 - 179944876 - 180007359 on NCBI Build GRCh38 |
Downstream | ubiquitin conjugating enzyme E2 E3 | UBE2E3 |
UBCH9, UbcM2 |
63.4kb | Sense | Chr. 2 - 180980381 - 181063427 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
