Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007678.hg.1 |
ENST00000468961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007678.hg.1 |
ENST00000468961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007678.hg.1 |
NR_104326.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007678.hg.1 |
NR_104326.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155491.hg.1 |
ENST00000468961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155491.hg.1 |
NR_104326.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155491.hg.1 |
ENST00000468961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155491.hg.1 |
NR_104326.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155491.hg.1 |
TC0300007678.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300155490.hg.1 |
ENST00000468961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155490.hg.1 |
NR_104326.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155490.hg.1 |
ENST00000468961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155490.hg.1 |
NR_104326.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155490.hg.1 |
TC0300007678.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007678.hg.1 |
ENST00000468961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007678.hg.1 |
ENST00000468961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007678.hg.1 |
NR_104326.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007678.hg.1 |
NR_104326.1 |
Refseq |