Assay Details
| Gene Symbol: SLAMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104400.1 | 3 - 4 | 1070 | 57 | ||
| GenBank mRNA | AK311666.1 | - | 2 - 3 | 330 | 57 | |
| BC144498.1 | - | 3 - 4 | 736 | 57 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CD84 molecule | CD84 |
LY9B, SLAMF5, hCD84, mCD84 |
-33.0kb | Sense | Chr. 1 - 160541094 - 160579516 on NCBI Build GRCh38 |
| Downstream | CD48 molecule | CD48 |
BCM1, BLAST, BLAST1, MEM-102, SLAMF2, hCD48, mCD48 |
43.8kb | Sense | Chr. 1 - 160678746 - 160711851 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SLAMF1
Genomic Map