Assay Details
| Gene Symbol: SLC25A44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104408.1 | 2 - 3 | 471 | 82 | ||
| GenBank mRNA | HY053859.1 | - | 2 - 3 | 303 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | semaphorin 4A | SEMA4A |
CORD10, RP35, SEMAB, SEMB |
-18.3kb | Sense | Chr. 1 - 156149673 - 156177751 on NCBI Build GRCh38 |
| Downstream | polyamine-modulated factor 1 | PMF1 | 5.0kb | Sense | Chr. 1 - 156212988 - 156240077 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SLC25A44
Genomic Map