Assay Details
| Gene Symbol: DACT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104425.1 | 1 - 2 | 178 | 61 | ||
| GenBank mRNA | BC111764.1 | - | 1 - 2 | 134 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929420 | LOC101929420 | -44.3kb | Anti-sense | Chr. 6 - 168242938 - 168262578 on NCBI Build GRCh38 | |
| Downstream | SPARC related modular calcium binding 2 | SMOC2 |
DTDP1, MST117, MSTP117, MSTP140, SMAP2, bA270C4A.1, bA37D8.1, dJ421D16.1 |
121.4kb | Anti-sense | Chr. 6 - 168441151 - 168667994 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC111764.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
epithelial cell morphogenesis
negative regulation of cell adhesion
regulation of Wnt signaling pathway
skin development
inner medullary collecting duct development
negative regulation of nodal signaling pathway
protein kinase C binding
beta-catenin binding
transcription factor binding
protein kinase A binding
delta-catenin binding
hematopoietic progenitor cell differentiation
epithelial cell morphogenesis
negative regulation of cell adhesion
regulation of Wnt signaling pathway
skin development
inner medullary collecting duct development
negative regulation of nodal signaling pathway
protein kinase C binding
beta-catenin binding
transcription factor binding
protein kinase A binding
delta-catenin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013962.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196236.hg.1 | TC0600013962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196234.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196234.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196234.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196234.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196234.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196234.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196234.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196234.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196234.hg.1 | TC0600013962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196247.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196247.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196247.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196247.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196247.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196247.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196247.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196247.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196247.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196247.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196247.hg.1 | TC0600013962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196235.hg.1 | TC0600013962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196245.hg.1 | TC0600013962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196243.hg.1 | TC0600013962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366795.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000366796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000607983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | ENST00000610183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286350.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_001286351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NM_214462.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NR_104425.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013962.hg.1 | NR_104425.1 | Refseq |

DACT2
Genomic Map