Assay Details
Gene Symbol: FRMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104455.1 | 12 - 13 | 1515 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
peptidylprolyl cis/trans isomerase, NIMA-interacting 4 pseudogene 1 |
PIN4P1 | -6.6kb | Sense | Chr. 15 - 43875806 - 43878171 on NCBI Build GRCh38 | |
Downstream | cancer susceptibility candidate 4 | CASC4 | H63 | 402.9kb | Anti-sense | Chr. 15 - 44288616 - 44415761 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
