Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000297423.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000517693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000518074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000519141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000519362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000519401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000521214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000521550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000522117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000524126.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000524141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000541342.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
NM_001080394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
NM_001282916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
NM_001282919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
NR_104581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000297423.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000517693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000518074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000519141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000519362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000519401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000521214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000521550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000522117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000524126.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000524141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
ENST00000541342.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
NM_001080394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
NM_001282916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
NM_001282919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
NR_104581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152051.hg.1 |
TC0800007529.hg.1 |
Clariom D TC |