Assay Details
| Gene Symbol: EXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104598.1 | 6 - 7 | 727 | 85 | ||
| XM_017014861.1 | XP_016870350.1 | 4 - 5 | 462 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NOTCH-regulated ankyrin repeat protein | NRARP | -71.2kb | Sense | Chr. 9 - 137299631 - 137302251 on NCBI Build GRCh38 | |
| Downstream | NADPH oxidase activator 1 | NOXA1 |
NY-CO-31, SDCCAG31, p51NOX |
48.6kb | Anti-sense | Chr. 9 - 137423372 - 137434406 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

EXD3
Genomic Map