Assay Details
Gene Symbol: LOC100506022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104622.2 | 1 - 2 | 76 | 100 | ||
GenBank mRNA | DA903593.1 | - | 1 - 2 | 76 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | splA/ryanodine receptor domain and SOCS box containing 1 | SPSB1 |
SSB-1, SSB1 |
-55.6kb | Anti-sense | Chr. 1 - 9292882 - 9369532 on NCBI Build GRCh38 |
Downstream | solute carrier family 25 member 33 | SLC25A33 |
BMSC-MCP, PNC1 |
110.5kb | Anti-sense | Chr. 1 - 9539469 - 9582773 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
