Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007139.hg.1 |
ENST00000503269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007139.hg.1 |
ENST00000503269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007139.hg.1 |
NR_104630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007139.hg.1 |
NR_104630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007139.hg.1 |
ENST00000503269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007139.hg.1 |
ENST00000503269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007139.hg.1 |
NR_104630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007139.hg.1 |
NR_104630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148321.hg.1 |
ENST00000503269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148321.hg.1 |
NR_104630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148321.hg.1 |
ENST00000503269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148321.hg.1 |
NR_104630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148321.hg.1 |
TC0500007139.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500148323.hg.1 |
ENST00000503269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148323.hg.1 |
NR_104630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148323.hg.1 |
ENST00000503269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148323.hg.1 |
NR_104630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148323.hg.1 |
TC0500007139.hg.1 |
Clariom D TC |