Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007219.hg.1 |
ENST00000507027.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007219.hg.1 |
ENST00000507027.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007219.hg.1 |
NR_104633.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007219.hg.1 |
NR_104633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148807.hg.1 |
ENST00000507027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148807.hg.1 |
NR_104633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148807.hg.1 |
ENST00000507027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148807.hg.1 |
NR_104633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148807.hg.1 |
TC0500007219.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500148808.hg.1 |
ENST00000507027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148808.hg.1 |
NR_104633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148808.hg.1 |
ENST00000507027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148808.hg.1 |
NR_104633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148808.hg.1 |
TC0500007219.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500007219.hg.1 |
ENST00000507027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007219.hg.1 |
ENST00000507027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007219.hg.1 |
NR_104633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007219.hg.1 |
NR_104633.1 |
Refseq |