Assay Details
Gene Symbol: GLIPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104639.1 | 4 - 5 | 540 | 96 | ||
GenBank mRNA | DA986727.1 | - | 4 - 5 | 341 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | reversion inducing cysteine rich protein with kazal motifs | RECK | ST15 | -26.4kb | Sense | Chr. 9 - 36036905 - 36124455 on NCBI Build GRCh38 |
Downstream | calicin | CCIN |
BTBD20, KBTBD14 |
8.8kb | Sense | Chr. 9 - 36169392 - 36171334 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
