Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
NR_104645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
NR_104645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
NR_104645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
NR_104645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
NR_104645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011280.hg.1 |
NR_104645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177372.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177372.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177372.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177372.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177372.hg.1 |
TC0X00011280.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00177373.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177373.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177373.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177373.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177373.hg.1 |
TC0X00011280.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000435394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
ENST00000609127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
NR_104644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
NR_104645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
NR_104645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
NR_104645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
NR_104645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
NR_104645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011280.hg.1 |
NR_104645.1 |
Refseq |