Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
ENST00000314957.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
ENST00000314957.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
ENST00000314957.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
ENST00000314957.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
ENST00000499871.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
ENST00000499871.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
ENST00000499871.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
ENST00000499871.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
ENST00000499871.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
ENST00000503152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
ENST00000503152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
ENST00000503152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034086.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034086.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034086.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034086.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034127.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034127.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034127.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034127.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034127.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_034127.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_104649.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_104649.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_104649.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_104649.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_104649.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_104650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_104650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_104650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_104651.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_104651.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010627.hg.1 |
NR_104651.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174190.hg.1 |
ENST00000503152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174190.hg.1 |
NR_104651.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174190.hg.1 |
ENST00000503152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174190.hg.1 |
NR_104651.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174190.hg.1 |
TC0500010627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500174193.hg.1 |
ENST00000503152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174193.hg.1 |
NR_104651.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174193.hg.1 |
ENST00000503152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174193.hg.1 |
NR_104651.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174193.hg.1 |
TC0500010627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
ENST00000314957.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
ENST00000499871.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
ENST00000503152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
NR_034086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
NR_104649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
NR_104650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
NR_104651.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
ENST00000314957.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
ENST00000499871.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
ENST00000503152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
NR_034086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
NR_104649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
NR_104650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
NR_104651.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174173.hg.1 |
TC0500010627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
ENST00000314957.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
ENST00000314957.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
ENST00000314957.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
ENST00000314957.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
ENST00000499871.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
ENST00000499871.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
ENST00000499871.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
ENST00000499871.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
ENST00000499871.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
ENST00000503152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
ENST00000503152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
ENST00000503152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034086.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_034127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_104649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_104649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_104649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_104649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_104649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_104650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_104650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_104650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_104651.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_104651.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010627.hg.1 |
NR_104651.1 |
Refseq |