Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
ENST00000513880.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
ENST00000513880.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
ENST00000513880.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013304.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007341.hg.1 |
ENST00000508923.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007341.hg.1 |
ENST00000508923.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007341.hg.1 |
ENST00000508923.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195536.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195536.hg.1 |
ENST00000513880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195536.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195536.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195536.hg.1 |
ENST00000513880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195536.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195536.hg.1 |
TC0500013304.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500195535.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195535.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195535.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195535.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195535.hg.1 |
TC0500013304.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500007341.hg.1 |
ENST00000508923.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007341.hg.1 |
ENST00000508923.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007341.hg.1 |
ENST00000508923.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195540.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195540.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195540.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195540.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195540.hg.1 |
TC0500013304.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500195538.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195538.hg.1 |
ENST00000513880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195538.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195538.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195538.hg.1 |
ENST00000513880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195538.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195538.hg.1 |
TC0500013304.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500195534.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195534.hg.1 |
ENST00000513880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195534.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195534.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195534.hg.1 |
ENST00000513880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195534.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195534.hg.1 |
TC0500013304.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500195541.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195541.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195541.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195541.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195541.hg.1 |
TC0500013304.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
ENST00000506534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
ENST00000513880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
ENST00000513880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
ENST00000513880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
NR_104653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013304.hg.1 |
NR_104653.1 |
Refseq |