Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000562888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000562888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000562888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010843.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184133.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184133.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184133.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184133.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184133.hg.1 |
TC1600010843.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000562888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000562888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000562888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010843.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184130.hg.1 |
ENST00000562888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184130.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184130.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184130.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184130.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184130.hg.1 |
ENST00000562888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184130.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184130.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184130.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184130.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184130.hg.1 |
TC1600010843.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600184129.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184129.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184129.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184129.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184129.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184129.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184129.hg.1 |
ENST00000569389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184129.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184129.hg.1 |
TC1600010843.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600184131.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184131.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184131.hg.1 |
ENST00000568137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184131.hg.1 |
NR_104657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184131.hg.1 |
TC1600010843.hg.1 |
Clariom D TC |