Assay Details
Gene Symbol: MAFTRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104663.1 | 2 - 3 | 157 | 68 | ||
GenBank mRNA | BX108785.1 | - | 2 - 3 | 157 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MAF bZIP transcription factor | MAF |
AYGRP, CCA4, CTRCT21, c-MAF |
-156.9kb | Sense | Chr. 16 - 79202624 - 79600725 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1229 | LINC01229 | 1.2kb | Anti-sense | Chr. 16 - 79770471 - 79801343 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185149.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185149.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185149.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185149.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185149.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185149.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185149.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185149.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185149.hg.1 | TC1600010955.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010955.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | ENST00000562921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | ENST00000566729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185146.hg.1 | TC1600010955.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600185148.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185148.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185148.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185148.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185148.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185148.hg.1 | ENST00000567993.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185148.hg.1 | ENST00000568389.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185148.hg.1 | NR_104663.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185148.hg.1 | TC1600010955.hg.1 | Clariom D TC |
