Assay Details
| Gene Symbol: LOC102467212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104670.1 | 4 - 5 | 404 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 5 open reading frame 30 | C5orf30 | -249.8kb | Anti-sense | Chr. 5 - 103258702 - 103278660 on NCBI Build GRCh38 | |
| Downstream | nudix hydrolase 12 | NUDT12 | 19.1kb | Sense | Chr. 5 - 103548855 - 103562805 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC102467212
Genomic Map