Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010943.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010943.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010943.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010943.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010943.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010943.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010943.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010943.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010943.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010943.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010943.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010943.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010943.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010943.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010943.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010943.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010943.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010943.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010943.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010943.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010943.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010943.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010943.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010943.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176315.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176315.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176315.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176315.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176315.hg.1 |
TC0500010943.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500176313.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176313.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176313.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176313.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176313.hg.1 |
TC0500010943.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500176314.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176314.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176314.hg.1 |
ENST00000508486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176314.hg.1 |
NR_105001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176314.hg.1 |
TC0500010943.hg.1 |
Clariom D TC |