Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013325.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195839.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195839.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195839.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195839.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195839.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195839.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195839.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195839.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195839.hg.1 |
TC0500013325.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500195840.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195840.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195840.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195840.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195840.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195840.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195840.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195840.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195840.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195840.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195840.hg.1 |
TC0500013325.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500195847.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195847.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195847.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195847.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195847.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195847.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195847.hg.1 |
TC0500013325.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500195838.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195838.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195838.hg.1 |
TC0500013325.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500195844.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195844.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195844.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195844.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195844.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195844.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195844.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195844.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195844.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195844.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195844.hg.1 |
TC0500013325.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500195841.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195841.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195841.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195841.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195841.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195841.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195841.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195841.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195841.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195841.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195841.hg.1 |
TC0500013325.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000506250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000506293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
ENST00000512573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
NR_105007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013325.hg.1 |
NR_105007.1 |
Refseq |