Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008164.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008164.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008164.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008164.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008164.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008164.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008164.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008164.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008164.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008164.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008164.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008164.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155989.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155989.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155989.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155989.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155989.hg.1 |
TC0500008164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500155991.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155991.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155991.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155991.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155991.hg.1 |
TC0500008164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500155993.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155993.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155993.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155993.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155993.hg.1 |
TC0500008164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500155995.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155995.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155995.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155995.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155995.hg.1 |
TC0500008164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008164.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008164.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008164.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008164.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008164.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008164.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008164.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008164.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008164.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008164.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008164.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008164.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155987.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155987.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155987.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155987.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155987.hg.1 |
TC0500008164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500155994.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155994.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155994.hg.1 |
ENST00000504012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155994.hg.1 |
NR_105028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155994.hg.1 |
TC0500008164.hg.1 |
Clariom D TC |