Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
ENST00000511256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
ENST00000511256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
NR_105049.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
NR_105049.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
NR_105049.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
NR_105050.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
NR_105050.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012094.hg.1 |
NR_105050.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183684.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183684.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183684.hg.1 |
ENST00000511256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183684.hg.1 |
NR_105049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183684.hg.1 |
NR_105050.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183684.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183684.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183684.hg.1 |
ENST00000511256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183684.hg.1 |
NR_105049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183684.hg.1 |
NR_105050.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183684.hg.1 |
TC0500012094.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500183687.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183687.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183687.hg.1 |
NR_105049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183687.hg.1 |
NR_105050.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183687.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183687.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183687.hg.1 |
NR_105049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183687.hg.1 |
NR_105050.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183687.hg.1 |
TC0500012094.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500183686.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183686.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183686.hg.1 |
ENST00000511256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183686.hg.1 |
NR_105049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183686.hg.1 |
NR_105050.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183686.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183686.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183686.hg.1 |
ENST00000511256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183686.hg.1 |
NR_105049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183686.hg.1 |
NR_105050.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183686.hg.1 |
TC0500012094.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
ENST00000507035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
ENST00000511256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
ENST00000511256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
NR_105049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
NR_105049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
NR_105049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
NR_105050.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
NR_105050.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012094.hg.1 |
NR_105050.1 |
Refseq |