Assay Details
| Gene Symbol: MIR7112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_107057.1 | 1 | 31 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | maestro heat like repeat family member 1 | MROH1 | HEATR7A | -733.0 | Anti-sense | Chr. 8 - 144148016 - 144261940 on NCBI Build GRCh38 |
| Downstream | scleraxis bHLH transcription factor | SCX |
SCXA, SCXB, bHLHa48 |
2.3kb | Anti-sense | Chr. 8 - 144265060 - 144268483 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0800012202.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000563210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000563210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000563210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000569712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | ENST00000616126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012202.hg.1 | NR_107057.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183458.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183458.hg.1 | ENST00000616126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183458.hg.1 | NR_107057.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183458.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183458.hg.1 | ENST00000616126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183458.hg.1 | NR_107057.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183458.hg.1 | TC0800012202.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000563210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000563210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000563210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000568812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000569712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | ENST00000616126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NM_015201.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012202.hg.1 | NR_107057.1 | Refseq |

MIR7112
Genomic Map