Assay Details
Gene Symbol: LINC01485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_108028.1 | 1 - 2 | 137 | 106 | ||
GenBank mRNA | CB162805.1 | - | 1 - 2 | 120 | 106 | |
DA640657.1 | - | 1 - 2 | 137 | 106 | ||
DA642776.1 | - | 1 - 2 | 101 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1484 | LINC01484 | -44.4kb | Sense | Chr. 5 - 173707599 - 173746209 on NCBI Build GRCh38 | |
Downstream | cytoplasmic polyadenylation element binding protein 4 | CPEB4 |
CPE-BP4, hCPEB-4 |
97.4kb | Anti-sense | Chr. 5 - 173888328 - 173960991 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000518675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000518675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197506.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197506.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197506.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197506.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197506.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197506.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197506.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197506.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197506.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197506.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197506.hg.1 | TC0500013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | ENST00000518675.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | ENST00000518675.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197508.hg.1 | TC0500013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500197496.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197496.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197496.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197496.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197496.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197496.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197496.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197496.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197496.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197496.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197496.hg.1 | TC0500013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500197497.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197497.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197497.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197497.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197497.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197497.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197497.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197497.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197497.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197497.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197497.hg.1 | TC0500013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500197505.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197505.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197505.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197505.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197505.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197505.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197505.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197505.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197505.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197505.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197505.hg.1 | TC0500013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500197495.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197495.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197495.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197495.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197495.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197495.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197495.hg.1 | TC0500013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500197509.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197509.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197509.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197509.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197509.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197509.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197509.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197509.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197509.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500197509.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500197509.hg.1 | TC0500013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000518675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000518675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000520324.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000522571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000522731.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | ENST00000523617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | NR_108028.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013405.hg.1 | NR_108028.1 | Refseq |
