Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007309.hg.1 |
ENST00000502457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007309.hg.1 |
ENST00000502457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007309.hg.1 |
ENST00000502457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007309.hg.1 |
NR_108034.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007309.hg.1 |
NR_108034.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007309.hg.1 |
NR_108034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500149424.hg.1 |
ENST00000502457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500149424.hg.1 |
NR_108034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500149424.hg.1 |
ENST00000502457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500149424.hg.1 |
NR_108034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500149424.hg.1 |
TC0500007309.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500007309.hg.1 |
ENST00000502457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007309.hg.1 |
ENST00000502457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007309.hg.1 |
ENST00000502457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007309.hg.1 |
NR_108034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007309.hg.1 |
NR_108034.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007309.hg.1 |
NR_108034.1 |
Refseq |