Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007713.hg.1 |
ENST00000432535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007713.hg.1 |
ENST00000432535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007713.hg.1 |
ENST00000432535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007713.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007713.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007713.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007713.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007713.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152404.hg.1 |
ENST00000432535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152404.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152404.hg.1 |
ENST00000432535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152404.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152404.hg.1 |
TC1000007713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000152408.hg.1 |
ENST00000432535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152408.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152408.hg.1 |
ENST00000432535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152408.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152408.hg.1 |
TC1000007713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000152411.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152411.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152411.hg.1 |
TC1000007713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000152403.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152403.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152403.hg.1 |
TC1000007713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000152407.hg.1 |
ENST00000432535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152407.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152407.hg.1 |
ENST00000432535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152407.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152407.hg.1 |
TC1000007713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007713.hg.1 |
ENST00000432535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007713.hg.1 |
ENST00000432535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007713.hg.1 |
ENST00000432535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007713.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007713.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007713.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007713.hg.1 |
NR_108046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007713.hg.1 |
NR_108046.1 |
Refseq |