Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
ENST00000454056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
ENST00000454056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
ENST00000454056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
ENST00000454056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166586.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166586.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166586.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166586.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166586.hg.1 |
TC1000009749.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000166585.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166585.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166585.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166585.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166585.hg.1 |
TC1000009749.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
ENST00000428165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
ENST00000454056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
ENST00000454056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
ENST00000454056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
ENST00000454056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009749.hg.1 |
NR_108058.1 |
Refseq |