Assay Details
| Gene Symbol: LOC100507642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108064.1 | 1 - 2 | 419 | 56 | ||
| GenBank mRNA | DB179823.1 | - | 1 - 2 | 379 | 56 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102723672 | LOC102723672 | -263.0 | Anti-sense | Chr. 7 - 144444 - 149438 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105375115 | LOC105375115 | 22.4kb | Sense | Chr. 7 - 174920 - 176013 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144085.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144085.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144085.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144085.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144085.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144085.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144085.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144085.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144085.hg.1 | TC0700006442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144092.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144092.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144092.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144092.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144092.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144092.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144092.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144092.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144092.hg.1 | TC0700006442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006442.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144093.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144093.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144093.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144093.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144093.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144093.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144093.hg.1 | TC0700006442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144087.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144087.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144087.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144087.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144087.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144087.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144087.hg.1 | TC0700006442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144094.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144094.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144094.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144094.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144094.hg.1 | TC0700006442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144095.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144095.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144095.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144095.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144095.hg.1 | TC0700006442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144096.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144096.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144096.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144096.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144096.hg.1 | TC0700006442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144084.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144084.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144084.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144084.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144084.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144084.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144084.hg.1 | TC0700006442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144091.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144091.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144091.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144091.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144091.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144091.hg.1 | ENST00000471299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144091.hg.1 | ENST00000479592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144091.hg.1 | ENST00000484550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144091.hg.1 | ENST00000492208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144091.hg.1 | NR_108064.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144091.hg.1 | TC0700006442.hg.1 | Clariom D TC |

LOC100507642
Genomic Map