Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011414.hg.1 |
ENST00000432246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011414.hg.1 |
ENST00000432246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011414.hg.1 |
ENST00000432938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011414.hg.1 |
ENST00000432938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011414.hg.1 |
ENST00000432938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011414.hg.1 |
NR_108070.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011414.hg.1 |
NR_108070.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011414.hg.1 |
NR_108070.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011414.hg.1 |
NR_108071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011414.hg.1 |
NR_108071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011414.hg.1 |
NR_108071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177490.hg.1 |
ENST00000432246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177490.hg.1 |
NR_108070.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177490.hg.1 |
ENST00000432246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177490.hg.1 |
NR_108070.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177490.hg.1 |
TC1000011414.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011414.hg.1 |
ENST00000432246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011414.hg.1 |
ENST00000432246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011414.hg.1 |
ENST00000432938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011414.hg.1 |
ENST00000432938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011414.hg.1 |
ENST00000432938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011414.hg.1 |
NR_108070.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011414.hg.1 |
NR_108070.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011414.hg.1 |
NR_108070.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011414.hg.1 |
NR_108071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011414.hg.1 |
NR_108071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011414.hg.1 |
NR_108071.1 |
Refseq |