Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009314.hg.1 |
ENST00000452933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009314.hg.1 |
ENST00000452933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009314.hg.1 |
ENST00000452933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009314.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009314.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009314.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009314.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009314.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162536.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162536.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162536.hg.1 |
TC1300009314.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300162537.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162537.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162537.hg.1 |
TC1300009314.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300162538.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162538.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162538.hg.1 |
TC1300009314.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300162540.hg.1 |
ENST00000452933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162540.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162540.hg.1 |
ENST00000452933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162540.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162540.hg.1 |
TC1300009314.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300162543.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162543.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162543.hg.1 |
TC1300009314.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009314.hg.1 |
ENST00000452933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009314.hg.1 |
ENST00000452933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009314.hg.1 |
ENST00000452933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009314.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009314.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009314.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009314.hg.1 |
NR_108076.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009314.hg.1 |
NR_108076.1 |
Refseq |