Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011430.hg.1 |
ENST00000436600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011430.hg.1 |
ENST00000436600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011430.hg.1 |
ENST00000436600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011430.hg.1 |
NR_108105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011430.hg.1 |
NR_108105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011430.hg.1 |
NR_108105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181139.hg.1 |
ENST00000436600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181139.hg.1 |
NR_108105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181139.hg.1 |
ENST00000436600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181139.hg.1 |
NR_108105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181139.hg.1 |
TC0700011430.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700181134.hg.1 |
ENST00000436600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181134.hg.1 |
NR_108105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181134.hg.1 |
ENST00000436600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181134.hg.1 |
NR_108105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181134.hg.1 |
TC0700011430.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700181140.hg.1 |
NR_108105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181140.hg.1 |
NR_108105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181140.hg.1 |
TC0700011430.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011430.hg.1 |
ENST00000436600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011430.hg.1 |
ENST00000436600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011430.hg.1 |
ENST00000436600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011430.hg.1 |
NR_108105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011430.hg.1 |
NR_108105.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011430.hg.1 |
NR_108105.2 |
Refseq |