Assay Details
| Gene Symbol: B4GALT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108108.1 | 3 - 4 | 523 | 146 | ||
| NR_108109.1 | 4 - 5 | 270 | 146 | |||
| GenBank mRNA | HY043002.1 | - | 3 - 4 | 192 | 146 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | beta-1,4-galactosyltransferase 1 | B4GALT1 |
B4GAL-T1, CDG2D, GGTB2, GT1, GTB, beta4Gal-T1 |
-11.5kb | Anti-sense | Chr. 9 - 33110641 - 33167391 on NCBI Build GRCh38 |
| Downstream |
serine peptidase inhibitor, Kazal type 4 |
SPINK4 |
HEL136, PEC-60, PEC60 |
60.2kb | Sense | Chr. 9 - 33240198 - 33248567 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147245.hg.1 | TC0900006966.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900147240.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147240.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147240.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147240.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147240.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147240.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147240.hg.1 | TC0900006966.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147244.hg.1 | TC0900006966.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147242.hg.1 | TC0900006966.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006966.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | ENST00000426270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147243.hg.1 | TC0900006966.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900147246.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147246.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147246.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147246.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147246.hg.1 | ENST00000442432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900147246.hg.1 | NR_108108.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147246.hg.1 | NR_108109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147246.hg.1 | NR_108110.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900147246.hg.1 | TC0900006966.hg.1 | Clariom D TC |

B4GALT1-AS1
Genomic Map