Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009924.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009924.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009924.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009924.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009924.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009924.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009924.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009924.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009924.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009924.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168084.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168084.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168084.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168084.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168084.hg.1 |
TC0900009924.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900168085.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168085.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168085.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168085.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168085.hg.1 |
TC0900009924.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900168082.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168082.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168082.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168082.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168082.hg.1 |
TC0900009924.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009924.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009924.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009924.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009924.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009924.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009924.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009924.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009924.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009924.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009924.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168081.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168081.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168081.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168081.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168081.hg.1 |
TC0900009924.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900168083.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168083.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168083.hg.1 |
ENST00000626300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168083.hg.1 |
NR_109756.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168083.hg.1 |
TC0900009924.hg.1 |
Clariom D TC |