Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011314.hg.1 |
ENST00000433546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011314.hg.1 |
ENST00000433546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011314.hg.1 |
ENST00000433546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011314.hg.1 |
NR_109805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011314.hg.1 |
NR_109805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011314.hg.1 |
NR_109805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000523747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000523747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000523747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
ENST00000523747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
NR_125431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
NR_125431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
NR_125431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
NR_125431.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006669.hg.1 |
NR_125431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177173.hg.1 |
ENST00000433546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177173.hg.1 |
NR_109805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177173.hg.1 |
ENST00000433546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177173.hg.1 |
NR_109805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177173.hg.1 |
TC0900011314.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000417333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000512942.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000523747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000523747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000523747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
ENST00000523747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
NR_125431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
NR_125431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
NR_125431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
NR_125431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006669.hg.1 |
NR_125431.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011314.hg.1 |
ENST00000433546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011314.hg.1 |
ENST00000433546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011314.hg.1 |
ENST00000433546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011314.hg.1 |
NR_109805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011314.hg.1 |
NR_109805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011314.hg.1 |
NR_109805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177175.hg.1 |
ENST00000433546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177175.hg.1 |
NR_109805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177175.hg.1 |
ENST00000433546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177175.hg.1 |
NR_109805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177175.hg.1 |
TC0900011314.hg.1 |
Clariom D TC |