Assay Details
| Gene Symbol: ATP11A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109811.1 | 1 | 517 | 126 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ATP11A upstream neighbor | ATP11AUN |
C13orf35, SMABLO1 |
-68.2kb | Anti-sense | Chr. 13 - 112647044 - 112684497 on NCBI Build GRCh38 |
| Downstream | MCF.2 cell line derived transforming sequence like | MCF2L |
ARHGEF14, DBS, OST |
139.6kb | Anti-sense | Chr. 13 - 112894378 - 113099739 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

ATP11A-AS1
Genomic Map