Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000432995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000432995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000432995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_046539.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_046539.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_046539.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_046539.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_051993.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_051993.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_051993.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_051994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_051994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_051994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_051994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_109838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_109838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_109838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_109838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_109838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_109839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_109839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_109839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007335.hg.1 |
NR_109839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000432995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000432995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000432995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
ENST00000435636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_046539.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_046539.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_046539.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_046539.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_051993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_051993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_051993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_051994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_051994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_051994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_051994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_109838.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_109838.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_109838.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_109838.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_109838.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_109839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_109839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_109839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007335.hg.1 |
NR_109839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
ENST00000432995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
NR_051993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
NR_051994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
NR_109839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
ENST00000422052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
ENST00000432995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
NR_051993.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
NR_051994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
NR_109839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149678.hg.1 |
TC1300007335.hg.1 |
Clariom D TC |