Assay Details
Gene Symbol: KCNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109845.1 | 1 - 2 | 222 | 101 | ||
GenBank mRNA | HY035404.1 | - | 1 - 2 | 225 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | potassium voltage-gated channel subfamily A member 2 | KCNA2 |
EIEE32, HBK5, HK4, HUKIV, KV1.2, MK2, NGK1, RBK2 |
-36.7kb | Sense | Chr. 1 - 110593580 - 110631536 on NCBI Build GRCh38 |
Downstream | CD53 molecule | CD53 |
MOX44, TSPAN25 |
197.7kb | Anti-sense | Chr. 1 - 110871199 - 110899936 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
