Assay Details
Gene Symbol: WDR86-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109857.1 | 5 - 6 | 675 | 59 | ||
GenBank mRNA | BX092283.1 | - | 5 - 6 | 675 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | WD repeat domain 86 | WDR86 | -2.0kb | Anti-sense | Chr. 7 - 151377974 - 151410705 on NCBI Build GRCh38 | |
Downstream | crystallin gamma N | CRYGN | 16.6kb | Anti-sense | Chr. 7 - 151428832 - 151448790 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000460148.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000460148.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000460148.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000466677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000466677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000466677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000466677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000466677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000466677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000460148.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000460148.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000460148.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000466677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000466677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000466677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000466677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000466677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000466677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000480632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034012.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_034013.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009703.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700169300.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700169300.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700169300.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700169300.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700169300.hg.1 | ENST00000489632.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700169300.hg.1 | NR_109857.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700169300.hg.1 | TC0700009703.hg.1 | Clariom D TC |
