Assay Details
Gene Symbol: LOC101929207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109860.1 | 2 - 3 | 193 | 75 | ||
GenBank mRNA | AI218423.1 | - | 1 - 2 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 654 | LINC00654 | -3.3kb | Sense | Chr. 20 - 5496067 - 5504596 on NCBI Build GRCh38 | |
Downstream | glycerophosphocholine phosphodiesterase 1 | GPCPD1 |
EDI3, GDE5, GDPD6, PREI4 |
36.1kb | Sense | Chr. 20 - 5544434 - 5611026 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
