Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000523605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000523605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000523605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000523605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
NR_109876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
NR_109876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009135.hg.1 |
NR_109876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008476.hg.1 |
ENST00000557499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008476.hg.1 |
ENST00000557499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008476.hg.1 |
ENST00000557499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000523605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000523605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000523605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000523605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
NR_109876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
NR_109876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009135.hg.1 |
NR_109876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008476.hg.1 |
ENST00000557499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008476.hg.1 |
ENST00000557499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008476.hg.1 |
ENST00000557499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000523605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
NR_109876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000523605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
NR_109876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163930.hg.1 |
TC0500009135.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000523605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
NR_109876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000518431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000523605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
ENST00000524295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
NR_109876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163932.hg.1 |
TC0500009135.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500163924.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163924.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163924.hg.1 |
NR_109876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163924.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163924.hg.1 |
ENST00000524034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163924.hg.1 |
NR_109876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163924.hg.1 |
TC0500009135.hg.1 |
Clariom D TC |