Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000508992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000508992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000508992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000514657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000514657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000514657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
ENST00000514657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181798.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181798.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181798.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181798.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181798.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181798.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181798.hg.1 |
TC0500011864.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000508992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000508992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000508992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000511194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000514657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000514657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000514657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
ENST00000514657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011864.hg.1 |
NR_109881.1 |
Refseq |