Assay Details
| Gene Symbol: LINC01430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109893.1 | 1 - 2 | 25 | 62 | ||
| GenBank mRNA | DB076868.1 | - | 1 - 2 | 25 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hepatocyte nuclear factor 4 alpha | HNF4A |
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14 |
-17.1kb | Anti-sense | Chr. 20 - 44355801 - 44432845 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1620 | LINC01620 |
C20orf62, dJ1013A22.3 |
11.2kb | Sense | Chr. 20 - 44451984 - 44465344 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LINC01430
Genomic Map