Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
ENST00000354611.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
ENST00000441832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
ENST00000452512.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
ENST00000483328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
ENST00000493181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
NM_001039693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
NR_004862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
NR_109905.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
NR_109906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
NR_109907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
ENST00000354611.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
ENST00000441832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
ENST00000452512.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
ENST00000483328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
ENST00000493181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
NM_001039693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
NR_004862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
NR_109905.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
NR_109906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
NR_109907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212768.hg.1 |
TC0200015377.hg.1 |
Clariom D TC |