Assay Details
Gene Symbol: SLC35B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109914.1 | 1 - 2 | 624 | 148 | ||
XM_017010914.1 | XP_016866403.1 | 1 - 2 | 399 | 148 | ||
GenBank mRNA | BX647208.1 | - | 1 - 2 | 558 | 148 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | EEF1E1-BLOC1S5 readthrough (NMD candidate) | EEF1E1-BLOC1S5 | EEF1E1-MUTED | -327.3kb | Sense | Chr. 6 - 8013567 - 8102595 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC100506207 | LOC100506207 | 56.0 | Anti-sense | Chr. 6 - 8435623 - 8785445 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_017010914.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
3'-phosphoadenosine 5'-phosphosulfate biosynthetic process
mitophagy in response to mitochondrial depolarization
xenophagy
3'-phospho-5'-adenylyl sulfate transmembrane transport
3'-phosphoadenosine 5'-phosphosulfate transmembrane transporter activity
positive regulation of defense response to virus by host
3'-phosphoadenosine 5'-phosphosulfate biosynthetic process
mitophagy in response to mitochondrial depolarization
xenophagy
3'-phospho-5'-adenylyl sulfate transmembrane transport
3'-phosphoadenosine 5'-phosphosulfate transmembrane transporter activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173143.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600173143.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173143.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173143.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173143.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173143.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600173143.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173143.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173143.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173143.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173143.hg.1 | TC0600010757.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | ENST00000379660.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_001142541.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NM_015948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010757.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173146.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173146.hg.1 | NR_109914.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600173146.hg.1 | TC0600010757.hg.1 | Clariom D TC |
