Assay Details
| Gene Symbol: KIAA0101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109934.1 | 1 - 2 | 242 | 109 | ||
| GenBank mRNA | DT220867.1 | - | 1 - 2 | 239 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | casein kinase 1 gamma 1 | CSNK1G1 | CK1gamma1 | -24.7kb | Sense | Chr. 15 - 64165517 - 64356243 on NCBI Build GRCh38 |
| Downstream | thyroid hormone receptor interactor 4 | TRIP4 |
ASC-1, ASC1, HsT17391, SMABF1, ZC2HC5 |
117.0 | Anti-sense | Chr. 15 - 64387804 - 64455303 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

KIAA0101
Genomic Map