Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010543.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173478.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173478.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173478.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173478.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173478.hg.1 |
TC0500010543.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500173488.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173488.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173488.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173488.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173488.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173488.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173488.hg.1 |
TC0500010543.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500173491.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173491.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173491.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173491.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173491.hg.1 |
TC0500010543.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010543.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173489.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173489.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173489.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173489.hg.1 |
ENST00000512519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173489.hg.1 |
ENST00000513480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173489.hg.1 |
NR_109951.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173489.hg.1 |
TC0500010543.hg.1 |
Clariom D TC |